United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
EW-11009-58
Ohaus Pioneer™ Analytical Balance, 210 G X 0.1 Mg (115 VAC). Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11009-58
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Analytical Balances
>
Ohaus Analytical Balances
>
Ohaus Pioneer Analytical Balances
Share
|
Ohaus Pioneer Analytical Balance, 210 G X 0.1 Mg (115 VAC)
EW-11009-58
$2,580.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
The draft shield removes completely for thorough cleaning of all components within seconds
Easy to clean analytical draft shield – panels and doors are removable for easy cleaning
Selectable environmental settings – three filter modes and adjustable zero tracking adjust balance sensitivity to environmental disturbances or application requirements
Basic weighing, parts counting, and percent weighing, plus 15 weighing units including one custom unit
Adjustable zero tracking is ideal for slow-filling applications where sensitivity is a must
The Pioneer’s draft shield removes completely for complete cleaning of all components within seconds. Periodic cleaning helps avoid sample contamination and extends the life of the balance. A standard in-use cover provides added protection against spillage and minimizes damage to the display and keypad. Three user-selectable environmental settings help the balance adapt to harsh environments and other disturbances for greater weighing accuracy. An up front level indicator eliminates the need to look behind the balance during the leveling process -- a quick glance at the up-front indicator helps ensure the balance is level prior to each use. An integral security bracket prevents unauthorized movement or theft. An RS-232 interface allows connection to a printer or PC for a record of balance ID, user and project ID, and time and date.
What's Included:
115 VAC Adapter, protective in-use cover, below balance weighing hook, and a draft shield
Specifications
Product Type
Analytical Balances
Auto shutoff
Yes
Calibration
External
Capacity (g)
210
Capacity (oz)
7
Housing dimensions
20 x 29 x 32 cm
Dimensions (" W)
7-3/4
Dimensions (" H)
11-1/4
Dimensions (" D)
12-1/2
Pan size (dia)
3-1/2" (8.9 cm)
Readability (g)
0.0001
Linearity (mg)
±0.3
Repeatability (mg)
±0.1
Weighing units
mg, g, lb, ct, oz, ozt, dwt, tical, tola, mommes, baht, grain, mesghal, Newton, taels
Stabilization time
3 seconds
Stability filters
three filter modes
Power (VAC)
115
Stability Indicator
Yes
Output
RS-232
Brand
Ohaus
Model
PA 214
Accessories
Qty
/
Price
Availability
Laboratory Mass Set, Set of 13 masses
EW-01035-00
at
$145.00 USD/SET of 1
In stock.
Add to Cart
Ohaus Compact Dot Matrix Printer
EW-11901-59
at
$549.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Ohaus Compact Dot Matrix Printer Paper For 11901-59
EW-11901-60
at
$10.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Ohaus Ink Ribbon For Compact Dot Matrix Printer 11901-59
EW-11901-61
at
$32.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Ohaus RS-232 Cable And Adapter For Compact Dot Matrix Printer 11901-59 f/moisture bal
EW-11901-63
at
$115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
ASTM Class 4 Mass, 200 g
EW-11953-27
at
$73.00 USD/EACH
Usually ships in 13 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title