United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
5 >
EW-11901-48
Ohaus SD Industrial Scale, 35 Kg X 0.02 Kg (115 VAC)
Click to enlarge
More Product Details
Specifications
Related products
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11901-48
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Industrial Scales
>
Ohaus Industrial Scales
>
Ohaus SD Industrial Shipping Scales
Share
|
Ohaus SD Shipping Scales, 35 Kg X 0.02 Kg (115 VAC)
EW-11901-48
$185.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Designed for shipping, receiving, and general industrial weighing applications
Flexible mountingremote indicator with coiled cable may be placed up to 9 feet away for eye-level mounting
Battery operable for greater portability auto shutoff feature helps extend battery life
These bench scales feature a durable painted steel platform, remote indicator with two tactile function keys, battery operation, and flexible mounting capabilities. Ideal for home office, mail room, shipping and receiving, and general industrial weighing.
What's Included:
AC adapter and table/wall mounting brackets
Specifications
Product Type
Shipping Scales
Auto shutoff
Yes
Calibration
External
Capacity (kg)
35
Capacity (lbs)
77
Display
4-digit, 7-segment LCD, 0.8" H
Platform size (L x W)
12-3/8" x 11" (31.4 x 27.9 cm) treaded
Readability (g)
20
Readability (lbs)
0.05
Linearity (g)
±20
Repeatability (g)
±20
Max count resolution
1:1750
Weighing units
kg, lb
Stabilization time
3 seconds or less
Power (VAC)
115
Battery
3 C batteries (not included)
Brand
Ohaus
Model
SD35
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Instruction Manual (4.8 MB)
Virtual Catalog (PDFs)
13-14 Catalog
More info
Ohaus SD Shipping Scale Data Sheet(470 KB)
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title