United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 17
of
18 >
EW-11010-71
Ohaus Scout® Pro Portable Balance, 6000 g/.1, 115 VAC. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11010-71
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Toploading Portable Balances
>
Ohaus Portable Balances
>
Ohaus Scout Pro Portable Balances
Share
|
Ohaus Scout Pro Portable Balance, 6000 g/.1, 115 VAC
EW-11010-71
$660.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Superior performance and easy two-button operation
Excellent shock resistance and overload protection
Software lock switch and integrated transport lock
Parts counting with auto optimization, % weighing, totalization, and display hold
Optional snap-in RS-232 and USB interfaces are easy to install
Scout Pro balances may be connected to a printer for convenient record-keeping.
For printer set up, order Thermal Printer 11901-47, and USB interface (11010-94 ) or RS232 interface (11010-92) separately. Add adapter 11901-58 for use with RS232 interface.
What's Included:
Weigh-below hook, AC adapter, calibration mass (200-, 400-, and 600g models)
Specifications
Product Type
Portable Balances
Calibration
External
Capacity (g)
6000
Capacity (oz)
211.6
Housing dimensions
19.1 x 5.4 x 21 cm
Dimensions (" W)
7-1/2
Dimensions (" H)
2-1/8
Dimensions (" D)
8-1/4
Pan size (L x W)
5-1/2" x 6-1/2" (14 x 16.5 cm)
Readability (g)
0.1
Linearity (g)
±0.01
Repeatability (g)
±0.01
Weighing units
g, kg, oz, lb, lb:oz, t, dwt
Power (VAC)
115
Battery
4 AA batteries (not included)
Stability Indicator
Yes
Output
RS-232 / USB / auxiliary
Brand
Ohaus
Model
SP6001
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
Ohaus security cable for Adventurer, Navigator, Scout and high capacity Explorer Pro balances
EW-11010-55
at
$57.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Ohaus RS-232 interface cable
EW-11010-92
at
$99.00 USD/EACH
In stock.
Add to Cart
Ohaus USB interface cable
EW-11010-94
at
$99.00 USD/EACH
In stock.
Add to Cart
Ohaus Compact Thermal Printer
EW-11901-47
at
$467.00 USD/EACH
In stock.
Add to Cart
Ohaus RS-232 Cable Adapter For Compact Thermal Printer 11901-47
EW-11901-58
at
$19.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer Balance Draft Sheild, Medium 15”W x 15-1/2”H x 16-3/4”D
EW-01019-04
at $138.00 USD/EACH
In stock.
Add to Cart
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Instruction Manual (1.61 MB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title