United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
7 >
EW-01012-00
Ohaus Triple-Beam Balance, 610 g x 0.1 g, Stainless Steel weighing plate
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-01012-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Mechanical Balances
>
Ohaus Triple-Beam Mechanical Balances
Share
|
Ohaus Triple-Beam Balance, 610 g x 0.1 g, Stainless Steel weighing plate
EW-01012-00
$213.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Classic mechanical balance with the durability and versatility to accommodate a range of lab applications
Spring-loaded zero adjust compensator
Magnetic damping speeds up weighing times
Select models feature a tare beam to accommodate containers up to 225 g
Dial-O-Gram® models come with convenient direct-reading dial calibration
Order weight set separately
Specifications
Product Type
Mechanical Balances
Balance type
triple-beam
Beam calibrations
front beam: 10 g x 0.1 g, middle beam: 500 g x 100 g, rear beam: 100 g x 10 g
Calibration
External
Capacity (g)
610
Capacity (kg)
0.61
Capacity (oz)
21.5
Capacity (lbs)
1.34
Capacity (with weight set)
2610 g
Housing dimensions
45.7 x 30.5 x 22.9 cm
Dimensions (" W)
18
Dimensions (" H)
12
Dimensions (" D)
9
Pan size (dia)
6" (15.2 cm)
Pan type
stainless steel
Readability (g)
0.1
Repeatability (g)
0.1
Weighing units
g
Brand
Ohaus
Model
760-00
Accessories
Qty
/
Price
Availability
Metric Attachment Weight Set 2x1000g, 1x500g
EW-01014-01
at
$45.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Ohaus Vinyl Dust Cover
EW-01021-00
at
$22.00 USD/EACH
Usually ships in 11 days.
Add to Cart
Ohaus Scoop, PP
EW-01329-00
at
$21.60 USD/EACH
Usually ships in 8 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title