United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
14 >
EW-11011-82
Ohaus Valor™ 3000 Xtreme Portion Control Scale, 2000g X 0.1g. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11011-82
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Food and Portion Scales
>
Ohaus Food Portion Scales
>
Ohaus Valor 3000 Xtreme Portion Control Scales
Share
|
Ohaus Valor 3000 Xtreme Portion Control Scale, 2000g X 0.1g
EW-11011-82
$557.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Extreme performance in an extremely rugged stainless steel design
No exposed screws or edges—perfect for food testing and service environments
All stainless steel design with shock and overload protection for tough environments
NSF certified, USDA-AMS accepted, and HACCP-certified
Washdown IP65-rated models may be rinsed clean with sprayed water
These compact industrial balances offer an extremely rugged design for long-term performance in tough environments. Valor is NSF certified, USDA-AMS accepted, and supports HACCP-certified systems. Washdown IP65-rated models may be rinsed clean with sprayed water. No exposed screws or edges make these balances perfect for food testing and service environments and offer easy cleanup. Ohaus’ classic SmarText™-driven interface includes a brilliant backlit display and the Yes/No menu navigation offers fast, user-intuitive operation. User-adjustable stability settings and resolutions allow the Valor 3000 series to operate accurately in drafty and unstable environments.
What's Included:
AC power adapter
Specifications
Product Type
Portion Control Balances
Calibration
External
Capacity (kg)
2
Capacity (lbs)
4.4
Housing dimensions
18.6 x 7.6 x 21.6 cm
Dimensions (" W)
7-5/16
Dimensions (" H)
3
Dimensions (" D)
8-1/2
Display
LCD
Pan size (L x W)
5-3/4" x 6-1/4" (14.6 x 15.9 cm)
Readability (g)
0.1
Linearity (g)
±0.1
Repeatability (g)
0.1
Weighing units
g, fractional or decimal oz, kg, lb, lb:oz, fluid oz
Power (VAC)
115
Battery
4 C batteries (not included)
Stability Indicator
Yes
Brand
Ohaus
Model
V31XH2
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, C, 4/pack
EW-09376-02
at
$12.55 USD/PACKAGE of 4
In stock.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Ohaus Valor 3000 Instruction Manual (1.2 MB)
Virtual Catalog (PDFs)
13-14 Catalog
More info
Ohaus Valor 3000 Data Sheet (215KB)
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title