United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
8 >
EW-11900-01
Ohaus Valor 5000 Portion Control Scale, 3 kg x 0.0001 kg / 6 lb x 0.002 lb
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11900-01
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Food and Portion Scales
>
Ohaus Food Portion Scales
>
Ohaus Valor 5000 Portion Control Scales
Share
|
Ohaus Valor 5000 Portion Control Scale, 3 kg x 0.0001 kg / 6 lb x 0.002 lb
EW-11900-01
$477.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
Rugged, multi-use scale designed for use in a wide variety of preparation and service applications
NSF certified, USDA accepted, and HACCP compliance
Spillproof front panel
Easy-to-clean smooth ABS plastic housing
Removable stainless steel pan is dishwasher safe
Manual, auto, or optional remote tare function
Checkweighing mode features three LED indicators and audible alarm
Percent weight function allows users to compare a sample as a percentage of a reference weight
Powered by AC or 120-hour rechargeable battery
NSF certified, USDA accepted, and HACCP compliant
What's Included:
Clear in-use cover, AC power adapter, and internal rechargeable battery
Specifications
Product Type
Portion Control Balances
Auto shutoff
Yes
Calibration
External
Capacity (kg)
3
Capacity (lbs)
6
Housing dimensions
30 x 22.3 x 18.5 cm
Dimensions (" W)
9
Dimensions (" H)
12
Dimensions (" D)
2-3/4
Display
Backlit blue LCD display
Display column height
0.75 inch / 19 mm
Platform dimensions
9.1 x 12 in / 228 x 305 mm
Pan size (L x W)
8-1/4" x 8-1/4" (20.9 x 20.9 cm)
Readability (g)
0.1
Readability (lbs)
0.0002
Linearity (g)
±0.1
Repeatability (g)
0.1
Weighing units
g, oz, kg, lb
Stabilization time
2 seconds
Power (VAC)
120
Battery
internal rechargeable
Low battery indicator
Yes
Languages
English
Brand
Ohaus
Model
V51PH3
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title