United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-04149-00
One-tier mills
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-04149-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
High-Capacity Laboratory Jar Mills
>
One-Tier Mills
Share
|
One-tier jar high-capacity laboratory jar mill; 13" roller, 115 VAC/60 Hz
EW-04149-00
$2,600.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Compact, space-saving design that's easy to operate
These laboratory mills incorporate all-welded steel frames, powerful roller chain drives, and pillow blocks with cartridge-type permanently sealed ball bearings for efficient operation.
Totally enclosed 1/4-hp DC motors and solid-state controls permit variable roller speed selection from 20 to 300 rpm for grinding a variety of wet and dry materials. The long-wearing, chemical-resistant rollers are self-centering and resistant to static buildup. Rollers easily adjust to accommodate a variety of jar diameters—no clamping or fastening required.
Choose a mill with either one or two tiers from the table below. One-tier mill 04149-10 accepts two or three jars at one time—grinds twice the amount of the other one-tier mills. Two-tier mills rotate several jars at one time and save horizontal space. Each tier accepts jars measuring from 2" (5.1 cm) to 10" (25.4 cm) in diameter.
What's Included:
the 115 VAC models include cord with standard plug; the 230 VAC model includes cord only. Mills require milling jars and grinding media; order separately on right.
Specifications
Product Type
Jar Mills
Description
one-tier
Motor
0.333 hp
Max load
60 lb/tier
Power (Hz)
60
Roller length
13 in
Dimensions
16 in L x 10.5 in W x 10.6 in H (40.6 x 26.7 x 26.9 cm)
Power (VAC)
115
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title