United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
25 >
Representative photo only
PAXT0010 Temperature Panel Meter Thermocouples T E J K R S B N C RTDs 100 Ohm Platinum 385 392 120 Ohm 11 36VDC 24VAC
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-65590-02
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-18
NIST-Traceable Calibration; Panel Meters
$75.00 USD/EACH/EACH
Usually ships in 10 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Panel Meters
>
Red Lion PAX Panel Meters
Share
|
PAXT0010, Temperature Panel Meter Thermocouples- T, E, J, K, R, S, B, N, C, RTDs- 100 Ohm Platinum 385/392, 120 Ohm, 11-36VDC/24VAC
EW-65590-02
$275.00 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Process, Voltage, Current, Temperature, And Strain Gauge Inputs
5-Digit 0.56'' Red Sunlight Readable Display
Variable Intensity Display
16 Point Scaling For Non-Linear Processprogrammable Function Keys/User Inputs
9 Digit Totalizer (Integrator) With Batching
Optional Custom Units Overlay W/Backlight
Our Setpoint Alarm Outputs (W/Option Card)
Communication And Bus Capabilities (W/Option Card)
Retransmitted Analog Output (W/Option Card)
Crimson Programming Software
NEMA 4X/IP65 Sealed Front Bezel
PART NUMBERS REPRESENT VARIOUS MODELS AND FEATURES. SOME PART NUMBERS ARE OUTPUT OR COMMUNICATIONS CARDS
The PAX
®
Analog Panel Meters offer many features and performance capabilities to suit a wide range of industrial applications. Available in five different models to handle various analog inputs, including DC Voltage/Current, AC Voltage/Current, Process, Temperature, and Strain gauge Inputs. The optional plug-in output cards allow the opportunity to configure the meter for present applications, while providing easy upgrades for future needs. The meters employ a bright 0.56'' LED display. The unit is available with a red sunlight readable or a standard green LED. The intensity of display can be adjusted from dark room applications up to sunlight readable, making it ideal for viewing in bright light applications.
The meters provide a MAX and MIN reading memory with programmable capture time. The capture time is used to prevent detection of false max or min readings which may occur during start-up or unusual process events.
The signal totalizer (integrator) can be used to compute a time-input product. This can be used to provide a readout of totalized flow, calculate service intervals of motors or pumps, etc. The totalizer can also accumulate batch weighing operations.
The meters have four setpoint outputs, implemented on Plug-in option cards. The Plug-in cards provide dual FORM-C relays (5A), quad FORM-A (3A), or either quad sinking or quad sourcing open collector logic outputs. The setpoint alarms can be configured to suit a variety of control and alarm requirements.
Specifications
Product Type
Temperature panel meters
Power (VDC)
11 to 36
CE Compliance
Yes
Power (VAC)
24
Input (thermocouple)
Yes
Input (RTD)
Yes
Display
5-digit, 0.56" (14.2 mm) Red LED, (-19999 TO 99999)
Operating conditions
0 to 50°C
Enclosure
NEMA 4X / IP65
Brand
Red Lion
Manufacturer number
PAXT0010
Model
PAXT0010
Accessories
Qty
/
Price
Availability
PAXCDS10, Plug-In Output Card Dual Setpoint Relay Output Card
EW-65590-11
at
$38.00 USD/EACH
In stock.
Add to Cart
PAXCDC20 Plug-In Communications Card RS232 Serial Communications Card
EW-65590-80
at
$49.00 USD/EACH
In stock.
Add to Cart
PAXCDC30 Plug-In Communications Card DeviceNet Communications Card
EW-65590-82
at
$122.00 USD/EACH
Usually ships in 4 days.
Add to Cart
PAXCDC50 Plug-In Communications Card Profi bus-DP Communications Card
EW-65590-85
at
$163.00 USD/EACH
Usually ships in 3 days.
Add to Cart
PAXCDC10 Plug-In Process Output Card (0-10VDC or 4-20mA) Analog Output Card
EW-65590-86
at
$75.00 USD/EACH
In stock.
Add to Cart
PAXCDS20 Plug-In Output Card Quad Setpoint Relay Output Card
EW-65590-88
at
$49.00 USD/EACH
Usually ships in 7 days.
Add to Cart
PAXCDS30 Plug-In Output Card Quad Setpoint Sinking Open Collector (NPN) Output Card
EW-65590-89
at
$38.00 USD/EACH
Usually ships in 4 days.
Add to Cart
PAXCDS40 Plug-In Output Card Quad Setpoint Sourcing Open Collector (PNP) Output Card
EW-65590-90
at
$38.00 USD/EACH
Usually ships in 4 days.
Add to Cart
PAXCDC10 Plug-In Communications Card RS485 Serial Communications Card
EW-68485-95
at
$49.00 USD/EACH
Usually ships in 4 days.
Add to Cart
PAXCDC1C Plug-In Communications Card Extended RS485 Communications Card
EW-68485-96
at
$49.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Label Kit Accessory
EW-68486-03
at
$11.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Panel Meters
EW-17090-18
at $75.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Process/Panel Meter Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title