Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

PTFE Film .005"thick x 12"W x 25ft rolls

EW-08277-15
$111.00 USD/ROLL

    Chemical resistant, temperature resistant, non-stick PTFE film protects benchtops, shelves, drawers, tables, trays, sinks, and other surfaces. Material is not porous, will not stain, and will not absorb odors or other contaminants. Reuseable film is easy to clean and easily cuts to size with scissors. Non-stick properties make this film ideal for use as a weighing sheet.
    Product Type Film
    Dimensions (ft/roll) 25
    Color Opaque
    Material PTFE
    Dimensions (" W) 12
    Thickness (mil) 1.3
    Brand Cole-Parmer
    Manufacturer number 15-5F-12
    Model 15-5F-12





    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title