United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-07090-62
PTFE-Diaphragm Pump Head
Click to enlarge
More Product Details
Specifications
Accessories
Replacement parts
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-07090-62
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Pump Heads
>
Masterflex L/S Pump Heads
>
L/S PTFE-Diaphragm Pump Heads
Share
|
PTFE-diaphragm pump head, 10 to 100 mL/min
EW-07090-62
$634.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Compatible with most Masterflex
®
L/S
®
drives
PTFE and glass wetted parts help maintain fluid purity.
Ideal for pumping through filters or into pressurized systems.
Check valve on inlet side of pump holds prime for up to two hours when pump is off.
Pumps include:
mounting tools and coupling hardware. (Note: Do not use these pump heads with Masterflex
®
drives requiring a long-shaft pump head.)
Service kits include:
one diaphragm, two check valve assemblies, two drive-tang boots an installation wrench, and instructions. Order external PTFE foot valves separately below to maintain prime.
Specifications
Product Type
Diaphragm Metering Pumps
Body material
PTFE
Dimensions
6-1/2"L x 3"W x 6"H
Max pressure continuous (psi)
75
Drive requirements
Minimum 1/20 hp
Brand
Masterflex
Manufacturer number
07090-62
Accessories
Qty
/
Price
Availability
Foot valve for 07090-62
EW-07090-65
at
$145.00 USD/EACH
In stock.
Add to Cart
Compression threaded adapter, PFA, 1/8" tubing OD x 1/8" female NPT
EW-31320-50
at
$27.50 USD/EACH
In stock.
Add to Cart
Compression threaded adapter, PFA, 1/4" tubing OD x 1/8" female NPT
EW-31320-52
at
$34.00 USD/EACH
Usually ships in 13 days or sooner.
Add to Cart
Replacement parts
Qty
/
Price
Availability
PFA Tubing, 1/16" ID x 1/8"OD, 25ft/pack
EW-06375-01
at $39.00 USD/PACKAGE
In stock.
Add to Cart
PFA Tubing, 5/32"ID x 1/4"OD, 25 ft/pack
EW-06375-02
at $94.00 USD/PACKAGE
Usually ships in 18 days or sooner.
Add to Cart
Service kit for 07090-62
EW-07090-63
at $277.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Masterflex® and the Human Heart: How One Pump is Supporting Research for the Other
Pump Head Torque Requirements
Pump Head Torque Requirements
Show All...
Occlusion: Questions and Answers
L/S® Pump Head Flow Performance
A Sweet Success: Famous Maple Syrup Producer Enhances Production with Masterflex® B/T® Variable-Speed Pump
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
The Ins and Outs of Metering Pumps
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Metering Pump Selection Guide
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
How to Read a Pump Curve
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title