United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 10
of
22 >
EW-36850-09
MiniMix Pictured
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36850-09
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Blenders and Paddle Blenders
>
Reciprocating Paddle Blenders
>
Interscience Bagmixer Paddle Lab Blender
Share
|
Paddle Blender; 5 To 100 mL, Variable Speed, Window Door, Removable Paddles
EW-36850-09
$4,350.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Adjustable paddles ensure quick and sterile blending of samples
Features click and clean system with removable paddles for easier cleaning
Designed for blending of small samples in food microbiology, clincial, and pharamaceutical applications
Variable blending speed (6-9 strokes per second)
Variable blending time (10-360 seconds or open running)
Clear window door allows visibility while blending
Security drip tray helps keep spills off the lab bench
The BagMixer® is the most effective lab blender for microbiological analysis. It guarantees optimal bacterial extraction of solid samples. Thanks to the unique adjustable paddle system, the blending chamber adapts itself perfectly to the nature and the size of the sample and works for any application.BagMixer® 100 MiniMix® is designed for the blending of small samples. Thanks to the adjustable paddles system, your blending is accurate and efficient.
The Click & Clean® system offers great user-comfort and
unequalled blending quality. The blending chamber is 100%
accessible for complete cleaning, due to the removable and
autoclavable paddles.
Applications:
Small samples
Food microbiology (BSE tests...)
Clinical, pharmaceutical
Veterinary (biopsy, organ tissue...)
Molecular extraction (proteins, lipids, DNA...)
Toxin research (HPLC, GCMS...)
Specifications
Product Type
Paddle Blenders
Description
MiniMix Click and Clean® paddle blender, window door
Power (VAC)
110/220
Power (Hz)
50/60
Blending speed
Variable, 3 to 12 strokes/sec.
Capacity
5 to 80 mL
Dimensions
20.3 x 20.3 x 36.8 cm
Door
Window
Dimensions (" W)
8
Dimensions (" H)
8
Dimensions (" D)
14-1/2
Brand
Interscience Laboratories Inc
Manufacturer number
013-110
Accessories
Qty
/
Price
Availability
Bagpage Paddle Blender Bags, 80ml With Full Filter, Sterile Packs Of 25 -- 500/pack
EW-36850-24
at
$289.00 USD/PACKAGE
Usually ships in 8 days.
Add to Cart
Bagpage Paddle Blender Bags, 100ml With Full Filter, Sterile Packs Of 25 -- 500/pack
EW-36850-25
at
$330.00 USD/PACKAGE
In stock.
Add to Cart
Baglight Paddle Blender Bags Without Filter, 100 mL, Sterile Packs Of 25 -- 500/pack
EW-36850-31
at
$77.00 USD/PACKAGE
Usually ships in 8 days.
Add to Cart
Bag Opener Stand For Holding Bags Size 80 And 100 mL, 1 Each
EW-36850-39
at
$125.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Storage Racks For 11 Bags 80 And 100ml, 1 Each
EW-36850-42
at
$175.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Closing Clips For Lab Blender Bags 80 And 100 mL
EW-36850-45
at
$135.00 USD/PACKAGE
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title