United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-90205-26
Panel-mount Remote Probe Thermometer
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-90205-26
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17003-00
NIST-Traceable Calibration; Bimetals or Pocket Digital Thermometers
$60.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Digital Thermometers
>
Digital Thermometers - General Purpose
>
Cole-Parmer Mini Digital Thermometers with Remote Probe
>
Cole-Parmer Panel-Mount Remote Probe Thermometer
Share
|
Panel-mount Remote Probe Thermometer
EW-90205-26
$23.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for measuring temperature from a distance.
Typical response time is 30 seconds for 99% of new reading
Thermometers have a 304 stainless steel penetration stem
All models have a NIST traceable certificate
Battery included
Panel-Mount Remote Probe Thermometer features a 10-ft flexible cord, a large easyview LCD, and records minimum and maximum readings over time. Versatile panel-mount module snaps into place or can be used as a stand-alone unit.Panel-mount Remote Probe Thermometer features a 10-ft flexible cord for measuring temperatures from a distance. Versatile panel-mount module snaps into place or can be used as a stand-alone unit. Model has an large easy-view LCD display, and records minimum and maximum readings over time.
Specifications
Product Type
Digital Indicator
Min temperature (º F)
-58
Min temperature (º C)
-50
Max temperature (º F)
572
Max temperature (º C)
300
Accuracy (º F)
±1.8°
Accuracy (º C)
±1.0°
Display
3.5 digit, 0.75 in
Probe (Remote)
Yes
Probe dimensions
2.75 in L
Resolution
0.1° F/C
Power
Battery (included)
Manufacturer number
4376CP
Model
4376CP
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Bimetals or Pocket Digital Thermometers
EW-17003-00
at $60.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
13-14 Catalog
Related technical articles (Roll over for summary)
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title