Parafilm Wrap PM998, 20" Wide; 50 Ft/Roll

WU-06720-25
$54.25 USD/ROLL

Are you a reseller or buying in bulk? Call us today.


  • Colorless, moisture proof plastic film molds to most surfaces
  • Ideal to wrap beakers and flasks
  • Select larger widths for covering trays
Multipurpose wrap is ideal accessory for any laboratory.
Film 06720-25 comes in a mailing tube.
Product Type Wrapping
Color Clear
Dimensions (" W) 20
Brand Parafilm
Model PM998
Manufacturer number PM998





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  • 13-14 Spanish Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title