United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
8 >
Representative photo only
Pelican Unbreakable Instrument Case 22 X 17 X 8 Interior
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-40460-88
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Handling and Storage
>
Instrument Cases
>
Pelican Unbreakable Instrument Cases
Share
|
Pelican Unbreakable Instrument Case, 22" X 17" X 8" Interior
EW-40460-88
$282.00 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
2
10%
$253.80/EACH
Usually ships in 13 days.
Qty
:
Add to Cart
Save to shopping list
Protect your valuable instrumentationcases are absolutely air- and water-tight
Customize your case with prescored “Pick-N-Pluck” foam
Cases are made of unbreakable, corrosion-proof polypropylene
Built-in O-ring keeps case air- and water-tight
(Instruments are not included)
All Pelican cases are constructed of high-impact polypropylene and feature ABS plastic latches. A ¼" thick neoprene O-ring guarantees a water- and dust-proof seal. Each case includes a "Pick-N-Pluck” foam linercustomize your case by simply removing pieces of foam.
Models 40460-92 and -94 also include adjustable handle and rolling wheels for easy transport.
Specifications
Interior dimensions
22"L x 17"W x 8"H
Product Type
Instrument case
Exterior Dimensions
24-"L x 19-"W x 9"H
Brand
Pelican
Manufacturer number
1600-000-110
Model
1600-000-110
Technical Resources
Related technical articles (Roll over for summary)
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title