United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 10
of
13 >
Representative photo only
Piston Metering Pump 1 4 stainless head 0 02 to 40 mL min range 1500 psi
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-73150-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Metering Pumps
>
Piston Pumps
>
Series Q Valveless Piston Pump Systems
>
Low-flow Piston Pump System
Share
|
Piston Metering Pump, 1/4" stainless head, 0.02 to 40 mL/min range, 1500 psi
EW-73150-50
$2,683.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Remote flow rate control via analog signal or RS232 interface
Optional liquid ends for expanded flow range
Smooth fluid delivery from intra RPM stepper motor control
Integrated piston wash
Extended flow ranges are possible by using up to three interchangeable liquid end assemblies. Software control of refill rates and compressibility compensation add to the versatility of the pumps and enhance pump performance. Precise metering is attained by a positive-displacement reciprocating piston assembly. The high-resolution stepper motor is used to maximize flow output and minimize pulsation.
A fully integrated piston wash system dramatically extends the life of the piston by rinsing away crystallized chemicals which lead to abrasion of the piston seal.
Choose pumps with an integrated pulse damper to further reduce pulsation. They also include the ability to monitor pressure, and set high and low limits.
Specifications
Product Type
Piston Metering Pumps
Wetted parts
316 SS body, Sapphire piston
Max pressure
1500 psi
Max viscosity (cp)
500
CE Compliance
Yes
Min flow rate (mL/min)
0.02
Max flow rate (mL/min)
40
Dimensions
4"W x 9"H x 9-1/2"D
Pump type
Piston
Repeatability
±0.3%
Port size
1/8" Swagelok inlet/outlet
Duty cycle
continuous
Max temperature (º C)
120
Max temperature (º F)
248
Technical Resources
Microfluidics brochure
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
The Ins and Outs of Metering Pumps
Metering Pump Selection Guide
How to Read a Pump Curve
Show All...
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title