log in or register
Plain Clipped-Corner Slides, 144/PK
EW-48520-01
$36.00 USD/PACKAGE
    Drastically reduce the chances of cutting your finger or tearing a glove when you use these slides. Clipped corners keep sample away from the edges of the slide—ideal for making blood smears. Slides measure 75 x 25 mm and are 1-mm thick. Available in one gross (144 slides).
    Product Type Microscope slides
    Quantity 144
    Brand Motic Instruments Inc
    Technical Resources
    Virtual Catalog (PDFs)
  • 11-12 Catalog
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title