Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Plate, 96 Well,ps,flat,100/cs

EW-01728-01
$304.00 USD/CASE of 100

Usually ships in 9 days.
Qty

  • Polystyrene plates meet the SBS standard for external dimensions—ensure compatibility with microplate and automation equipment
  • Round wells optimize coating and washing
  • Nontreated for solution-based homogeneous assays
  • Clear plates are ideal for colorimetric and storage applications
  • Clear, high-grade polystyrene for research
  • Formulated for uniform binding, high optical clarity, and low background absorption
  • Medium-binding plates offer a binding capacity of 250 ng of mouse IgG/cm2
Solid Plates come in clear, white, and black with either a nontreated, high-binding, or cell culture surface.
Clear-Bottom Plates optimize optical clarity and facilitate cell detection. Available in black or white with either a nontreated or cell culture surface.
Product Type Microplates
Sterile Yes
Surface treatment Untreated
Color Clear
Well shape Flat
Well volume (mL) 0.36
Lid Yes
Well format 96 well
Qty/sleeve 20
Material Polystyrene
Brand Corning
Manufacturer number 3370

Accessories
Qty/Price
Availability

at $214.00 USD/CASE of 100

Call 1-888-358-4705 for availability.

at $80.00 USD/CASE of 50

Call 1-888-358-4705 for availability.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title