United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-04186-00
Plate mill, with standard motor, 115 VAC, 60 Hz
Click to enlarge
More Product Details
Specifications
Accessories
Reconditioned & Like New
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04186-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Mills
>
Plate mills
Share
|
Plate mill, with standard motor, 115 VAC, 60 Hz
EW-04186-00
$2,050.00 USD/EACH
Usually ships in 15 days.
Qty
:
Add to Cart
Save to shopping list
Heavy-duty plate mills provide clog and heat-resistant operation
Low output speed makes mill ideal for grinding hard and dry materials
Grinding unit disassembles without tools for easy cleaning
Use this heavy-duty plate mill for a variety of grinding applications: food, pharmaceutical, chemical, soil testing, and ore and coal sampling. Grinding unit is constructed of cast iron and coated with lead-free tin for food handling applications. Adjust the thumbscrew on the grinding unit to change the plate gap from 1/4" open to nearly closed. Includes grinding plates for fine grinding; for coarse grinding, order optional coarse grinding plate set 04146-53 separately. Grinding plates and feeds are made of durable iron alloy. Use the tooth feed (included) for dry materials and the worm feed (optional) for wet or oily materials.
What's Included:
fine grinding plate, tooth feed, 8-ft (2.4-m) cord with standard U.S. plug (115 VAC) or European plug (230 VAC), and inline on/off switch. Order replacement plates, replacement tooth feed, and optional worm feed separately on right.
Specifications
Product Type
Plate Mills
Capacity
50 cu in.
Speed
89 rpm
Motor
0.333 hp
Final Particle Size
75 to 100 mesh
Power (Hz)
60
Dimensions
25 in L x 8 in W x 12 in H
Power
115 VAC, 60 Hz
Power (VAC)
230
Accessories
Qty
/
Price
Availability
Replacement fine grinding plate set
EW-04186-51
at
$115.00 USD/SET
Usually ships in 6 days.
Add to Cart
Coarse grinding plate set
EW-04186-53
at
$95.00 USD/SET
Usually ships in 6 days.
Add to Cart
Replacement tooth feed, for dry material
EW-04186-54
at
$116.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Worm feed, for wet or oily material
EW-04186-56
at
$116.00 USD/EACH
Usually ships in 11 days.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
RECOND 04186-00
EW
-
00260MQ
at
$1,307.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Smart Mixing
Mixer Selection Guide
Positioning a Mixer in a Cylindrical Vessel
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title