United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-51503-50
Platform for Cole-Parmer
®
High-Capacity Shakers
Representative photo only
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-51503-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Share
|
Replacement Accessories
-
Platform for Cole-Parmer
®
High-Capacity Shakers
EW-51503-50
$425.00 USD
(75% off)
$106.25 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Specifications
Product Type
Accessory
Brand
Cole-Parmer
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Job Title
|
Category
|
Brand
|
Part Number
|
Price
View product categories by:
Popularity
|
Favorites
|
Job Title
|
Price
|
Part Number
View popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Job Title
View reviews by:
Popularity
|
Product Title
|
Best Rated