United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-12105-80
PolyScience Coliform Testing Circulating Baths, 120V, 60Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-12105-80
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Specialty Baths
>
Coliform Baths
>
PolyScience Coliform Testing Circulating Baths
Share
|
PolyScience Coliform Testing Circulating Baths, 120V, 60Hz
EW-12105-80
$2,006.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Controller preset at 44.5°C complies with EPA testing regulations for fecal coliform contamination testing
Large backlit digital display can be seen from across most rooms
Integral re-circulating pump ensures uniform bath temperature
Hinged gabled see-through cover
For applications such as Standard Method Coliform Testing, general bacteriological testing, incubation, sample tempering, and routine laboratory use. This circulating bath is designed specifically for fecal coliform contamination testing, which is an indicator of E. Coli microorganisms and other pathogens presence. It is preset at 44.5°C temperature to comply with the EPA Total Coliform Rule (TCR) requirement.
Designed specifically for coliform testing, this lab-friendly circulating bath features the digital controller as shown on page 139 for the PolyScience
®
immersion circulators. This highly intuitive, easy-to-use controller makes it simple to set commonly used coliform test temperatures, such as 44.5, 45.5, 41.5, and 35.0°C, or other temperatures from ambient +10 to 135°C. Bath and set point temperature are simultaneously displayed on a large 2.12 x 3 inch LCD readout.
For operational convenience, this bath is equipped with a tilting, see-through polycarbonate cover which drains back into the bath when opened. The lid is gabled to allow for taller vessels. The port in the top deck to accommodate an optional tap-water cooling coil, and an integral 1-speed (simplex) pump which can be used to circulate the bath liquid to other instruments using a closed-loop configuration. For added safety and durability, the bath features a precision molded phenolic top deck that remains cooler at high temperatures and is easily cleaned and disinfected. All wetted components are corrosion-resistant stainless steel and high-performance polymer.
What's Included:
Bath cover, perforated stainless steel sample tray, 6-ft power cord and instruction manual.
Specifications
Product Type
Water Bath
Bath type
Recirculating Water Bath
Capacity (Liters)
4.5
Min temperature (º C)
Ambient
Max temperature (º C)
135
Temp stability
±0.07
Working area (" W)
11
Working area (" L)
8-5/8
Working area (" D)
8
Heater wattage
1100
Pump type
Force
Max flow rate (Liters/min)
8.9
Power (VAC)
120
Power (Hz)
60
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title