United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
PolyScience Constant Temperature Viscosity Bath 17 L 240 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-98935-24
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Physical Property Testing
>
Viscometers
>
Viscosity Baths
>
PolyScience Constant-Temperature Viscosity Baths
Share
|
PolyScience Constant-Temperature Viscosity Bath, 17 L, 240 VAC
EW-98935-24
$2,838.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
An economical alternative for kinematic viscosity measurement
These kinematic viscosity baths meets ASTM D-445 specifications for measuring petroleum, lubricants, polymer coatings, food, paint, and other liquids. Advanced PID controls maintain precise temperatures and quick responsiveness under changing heat load conditions. The integral recirculating pump ensures uniform temperature. Large, backlit digital display shows both set and actual values. Other features include Celsius/Fahrenheit selection, high/low temperature limits, chemical- and thermal-resistant top plate, and transparent tank for excellent visibility.
What's Included:
tank, top plate, circulator, covers for holes, and power cord.
Specifications
Product Type
Viscosity Bath
Min temperature (º C)
Ambient
Max temperature (º C)
135
Temp control
PID
Temp stability
±0.07°C
Temp setting
Digital
Power (VAC)
240
Display
LCD
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
L'affaire est ketchup? Pas si simple que ça...les mesures de viscosité
In the Thick of Things: Measuring Viscosity
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Show All...
Viscous Fluid Behavior
Equipping a Laboratory: A Study in Customer Service Excellence
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title