United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
24 >
EW-12119-04
PolyScience
®
Performance Programmable 7L Heating Bath Circulator 120V/60Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-12119-04
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Heated Circulators
>
PolyScience Heated Circulators
>
PolyScience Heating Circulating Baths
Share
|
PolyScience Performance Programmable 7L Heating Bath Circulator 120V/60Hz
EW-12119-04
$3,537.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Extra-large backlit digital display can be easily seen from across a room
Waterproof control panel swivels 180°, allowing viewing at almost any angle
Unique LidDock
®
system stores the lid in open position for easy sample placement and drains condensation back into reservoir
Color touch-screen models feature programmability and computer communication
Available in multiple control level options and a variety of tank sizes, PolyScience’s next-generation baths maintain precise temperature control for virtually any laboratory or process application. Advanced PID algorithms increase heat responsiveness, eliminate set point overshoot, and ensure a constant temperature. The patented WhisperCool™ environmental control system features part-load sound levels below that of conventional speech at a distance of two meters. The chemical resistant DuraTop™ top plate remains cool at high temperatures and is easily cleaned and disinfected. Variable-speed pump keeps the bath liquid at a uniform temperature. Rear inlet/outlet ports also allow use in external process applications.
Basic and Standard Digital Controllers
are manufactured with the EasyView™ LCD measuring 21⁄8" x 3" (5.4 x 7.6 cm). The Basic controller reaches a maximum temperature of 135°C and has a single-speed pump. The Standard Digital controller reaches up to 170°C and has a two-speed pump.
Performance Digital Controller
utilizes SmartTouch™ color touchscreen display and controls. These SmartTouch models are the ultimate in icon-driven, user friendly operation! Simply touch the screen to scroll through the menus, functions, and options—even when wearing gloves. This touch-screen digital display allows easy monitoring of both the set value and actual value. Touch and scroll operation with visual prompts make setup and operational changes simple and straightforward. View screen text in 11 languages. Data trending is visibly tracked on the display.
Performance Programmable Digital Controller
includes all the Performance Digital controller features and adds unlimited time/temperature programmability.
What's Included:
Bath cover and lid, power cord, drain valve, fittings, plumbing tape, clamps, drain tubing, and instruction manual. Performance Programmable models also include communication cables and memory stick
Specifications
Product Type
Water Bath
Bath type
Heated Water Bath
Capacity (Liters)
7
Min temperature (º C)
Ambient
Max temperature (º C)
200
Temp control
PID
Temp display
SmartTouch™ Color LCD 4.3 in
Temp sensor
RTD, Pt 1000 Ohms
Temp stability
±0.005°C
High-temperature cutoff
Yes
Low-level cutoff
Yes
Working area (" W)
5-5/8
Working area (" H)
5-1/2
Working area (" L)
6-3/16
Heater wattage
1100
Pump type
Force and Suction
Control type
Performance digital
Max pump pressure (psi)
4
Max pump pressure (mbar)
275
Max flow rate (Liters/min)
16
Power (VAC)
120
Power (Hz)
60
Power (Amps)
10
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title