United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
Representative photo only
Polypropylene fittings for 1 16 tubing 1 4 28 UNF M pack of 10
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-06471-58
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Fittings
>
Chromatography fittings
>
Couplings and tube end fittings
>
Inert Tube End Fittings
Share
|
Polypropylene fittings for 1/16" tubing, 1/4-28 UNF(M), pack of 10
EW-06471-58
$20.00 USD/PACKAGE
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for use with solenoid valves
No tools required for assembly to tubing!
Use these fittings with tubing ranging from 1 to 11 mm OD. All standard fittings except models 06473-07 and -08 come with polypropylene (PP) caps, two large VITON O-rings per cap to connect 2 to 4 mm OD tubing, one small VITON O-ring per cap (use together with large O-rings) to connect 1 to 2 mm OD tubing, and one PTFE cone per cap to make chemically inert connections for 1/16" OD tubing or to create zero dead volume fittings for connection to high-pressure fittings (sold below).
Standard fittings 06473-07 and -08 include one fluoroelastomer O-ring per PTFE cap. All chemically inert fittings come with TEFZEL
®
caps and 1/16" tube OD PTFE cones for improved chemical resistance. All O-rings are made of fluoroelastomer. Max fluid pressure (up to 50 psi) will vary depending on tubing material and size.These tube end fittings with inverted cones provide a flangeless fitting that is economical and quick to assemble. Just slip the nut over the tubing, fit the cone to the end of tubing, and screw into the port. The inverted cone sits in a recess in the fitting to allow for maximum thread engagement. The fitting will stay sealed when finger tight, even in shallow ports such as solenoid valves.
Specifications
Fitting body material: PTFE
Cone material: Tefzel
®
Nut material: glass-filled polypropylene
Sealing pressure: rated to 500 psi for 1/16" OD tubing, 250 psi for 1/8" OD tubing
Specifications
Product Type
Chromatography fittings
Material
Polypropylene
Quantity
10
Brand
Diba
Manufacturer number
008NC16-YC5U
Accessories
Qty
/
Price
Availability
Tefzel Cones, 1/16", 10 per pack
EW-06471-56
at
$20.00 USD/PACKAGE
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Sterilizing Fittings: The Critical Factors
Fittings Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title