United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-99561-25
PORTABLE JAR TESTER, 4 POSITIONS
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-99561-25
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Turbidity Meters and Turbidity Testing Products
>
Flocculation Testers
>
Orbeco-Hellige Jar Testers
Share
|
Portable Jar Tester, Portable, 4 Positions
EW-99561-25
$1,820.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
ASTM D203S compliant method
Single motor drives all positions at a consistent speed
Choose from laboratory or portable models
Available with 4 or 6 positions
Jar testers are designed to test the efficiency of flocculation or preceipitation reagents. The single motor drives all positions at a constant stirring speed. Stirring rod shafts can individually be stopped and height can be adjusted while the system is operating. Choose from laboratory or portable tester models.
Laboratory Jar Testers are available with either four or six positions with a digital display that shows stirring speed and time. Stirring speed is programmable from 10 to 300 rpm, and time from 1 to 999 minutes or 0 to 99 hours. Featuring an illuminated back panel for enhanced visibility. Laboratory units require standard 2-L beakers (order separately,34512-13 4/pk).
Portable Jar Tester is available in a space saving 4-position model with stirring positions around a central lamp. Set stirring speed mechanically to 20, 40, 50, 100 or 120 rpm, and set time for 1 to 30 minutes. Central lamp makes it easier to observe flocculation process. Portable tester requires 1-L beakers (order separately, 34512-11 6/pk).
Specifications
Product Type
Jar Tester
Meter style
Portable
Dimensions (" W)
12.5
Dimensions (" H)
6.6666666667
Dimensions (" L)
9.6666666667
Accuracy (Dissolved Oxygen)
±0.2mg/L from 0 to 20 mg/L; ±0.5 mg/L from 50 to 50 mg/L
Display
Analog
Power
AC Adapter 100/240 VAC
CE Compliance
Yes
Brand
Orbeco-Hellige
Model
ET 730
Accessories
Qty
/
Price
Availability
Pyrex Brand 1003 Griffin Heavy-Duty Beaker, 1000 mL
EW-34512-11
at
$153.00 USD/PACKAGE of 6
In stock.
Add to Cart
Technical Resources
Orbeco Portable Jar Tester
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Good Laboratory Practices for Taking Turbidity Measurements
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title