dummy
EW-68974-12
Call 1-888-358-4705 for availability.
Qty:
- Designed to calibrate pressure transmitters from vacuum to 300 psi
- Low-pressure models have accuracy of ±0.05% of span and ranges of ±1" H2O to 60"H2
- High-pressure models have accuracy of ±0.025% full-scale
- Choose from documenting and non-documenting calibrators in low or high pressure ranges
High-pressure calibrators include built-in pump for ease of pressure generation, and the volume adjuster for fine pressure control. Low-pressure calibrator includes a built-in dual piston volume adjuster for course/fine pressure settings. The pressure relief valve removes trapped air without the need to disconnect the device under test. Rugged NEMA 12 (IP54) case with elastomer keypad is designed for industrial or field use. Measure pressure in 25 different units including a user-definable scale. Dual display indicates current, voltage, temperature, and minimum/maximum readings. The TASK menu provides options for pressure transmitter calibration, pressure switch testing, leak testing, loop testing, and fault finding. To aid calibration and testing, calibrators supply 10 VDC, 24 VDC, and up to 24 mA. Models 68974-12 to -18 and -48 to -58 store up to 10,000 readings and send the data to a computer via a RS-232 port. Models 68974-40 to -46 and -60 to 70 feature two-way PC communication for transferring procedures and results, as well as error analysis with pass/fail status. Procedures downloaded from a PC automatically configure the calibrators to pre-defined calibration and test routines. Optional software is required. Order seperately below.
The Low-Pressure units are designed on-site calibrateion of very low pressure sensors. At low differential pressures, even connecting the device under test can induce significant pressure. To compensate for this, an “auto zero” facility is provided which uses an internal solenoid valve to equalize the differential pressure prior to zero correction.
| Product Type |
Pressure/Vacuum Calibrators |
| Description |
DPI 610 Calibrator |
| Range |
-14.7 to 100 psig measures approximately 30 in of Hg vacuum |
| Resolution |
0.01 |
| Media compatibility |
liquids or gases compatible with stainless steel |
| Accuracy |
Pressure: ±0.025% full-scale Current: ±0.05 of reading ±0.004% full-scale Voltage: ±0.05 of reading ±0.004% full-scale Compensated temperature: 15 to 105°F (10 to 40°C) |
| Dimensions |
11.875 in L x 6.75 in W x 5.5 in H |
| Power |
six 1.5 V size C NiCd batteries (included) |
| Brand |
Druck |
at $1,053.00 USD/EACH
Call 1-888-358-4705 for availability.
at $159.00 USD/EACH
Call 1-888-358-4705 for availability.
at $125.00 USD/EACH
Usually ships in 13 days.
at $103.00 USD/EACH
Usually ships in 13 days.
at $63.00 USD/EACH
Usually ships in 14 days.
at $1,256.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,006.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,006.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,006.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,006.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,006.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,006.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,256.00 USD/EACH
In stock.
at $1,256.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,256.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,517.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,256.00 USD/EACH
Call 1-888-358-4705 for availability.
at $135.00 USD/EACH
Usually ships in 14 days.
at $1,517.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,256.00 USD/EACH
Call 1-888-358-4705 for availability.
at $425.00 USD/EACH
Usually ships in 10 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.