dummy
EW-33703-30
Usually ships in 8 days.
Qty:
- Two self-supporting extractor arms are ideal for two-station setups
These air cleaning systems capture and remove smoke, dust, mist, and fumes before they enter the work area, then recirculate clean air back into the workplace. The systems are self-contained, with two 3"" dia self-supporting extractor arms made of heat-, chemical-, and abrasion-resistant polypropylene; end of arms flare up to 4 1/2"" dia. Ideal for two-station setups, or use the included inlet cap for single-arm use. Choose from bench top or portable system, then complete your system with filters sold separately.
Recommended for:
-Lab fumes and dust
-Clean room applications
-Soldering
-Small grinding applications
-Light welding
-Cleaning with solvents
-Art restoration & conservationPortable Air Cleaning System can be used as a two- or three-stage filtration system. The cleaner is mounted to a portable cabinet.
| Product Type |
Fume extractor systems |
| Exterior Dimensions (" W) |
20.5 |
| Exterior Dimensions (" H) |
36.8 |
| Exterior Dimensions (" D) |
13 |
| Power (VAC) |
115 |
| Power (Hz) |
60 |
| Noise level (dB) |
64 |
at $129.00 USD/EACH
Usually ships in 8 days.
at $237.00 USD/EACH
Usually ships in 10 days.
at $333.00 USD/EACH
Usually ships in 5 days.
at $494.00 USD/EACH
Usually ships in 5 days.
at $166.00 USD/EACH
Usually ships in 7 days.
at $87.00 USD/EACH
Usually ships in 5 days.
at $34.00 USD/EACH
Usually ships in 5 days.
at $46.00 USD/EACH
Usually ships in 5 days.
at $206.00 USD/EACH
Usually ships in 6 days.
at $39.00 USD/PACKAGE of 6
Usually ships in 11 days.
Technical Resources
Still have questions?
Contact Our Experts
for FREE product
application selection