United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-68604-00
Portable vacuum meter
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68604-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17040-12
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 0.75% or < 0.75%)
$140.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Digital and Analog Manometers
>
Portable Vacuum Meter
Share
|
Portable vacuum meter
EW-68604-00
$535.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Measure liquid, air, oil, and gas
Eight units of measure in easy-to-read display
Detachable heavy-duty probe
Auto shut-off
This cost-effective vacuum meter is ideal for vacuum measurement applications such as pneumatic, high heat, hydraulic, refrigeration, HVAC, and food processing. It features overload protection up to 2000 mbar max, built-in low battery indicator, and optional RS-232 serial interface and software for downloading data to a PC. Weighing less than one pound, meter records maximum and minimums with recall and a data hold function for storing desired value on display.
Specifications
Product Type
Vacuum gauges/controllers
Media compatibility
liquid, air, oil, gas
Accuracy
±1% full-scale
Unit of Measurement
mbar, psi, mm Hg, " Hg, kPa, Pa, torr, micron
Resolution
0.01 psi
Output
RS-232
Range
1 to 21.75 psi
Process connection
1/4" NPT(M)
Power
one 9 V battery (included)
Display
4-digit LCD, 5/8" (1.6 cm) H
Accessories
Qty
/
Price
Availability
RS-232 cable, cable rs232
EW-68601-90
at
$29.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Graphical software
EW-68604-50
at
$58.00 USD/EACH
In stock.
Add to Cart
Water-resistant pouch
EW-94461-25
at
$15.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 0.75% or < 0.75%)
EW-17040-12
at $140.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Anemometers
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Show All...
Pressure Unit Conversion Constants
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title