United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
12 >
Representative photo only
Precision ASTM Class 1 calibration mass set with an ISO IEC 17025 2005 accredited certificate 200 g to 1 mg
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11957-53
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17109-34
Calibration Service For mass Set With Maximum Value Between 100 To 200 G; Astm 1-3
$368.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Calibration Masses
>
Troemner Calibration Masses
>
Troemner Stainless Steel Precision Analytical Masses (UltraClass through Class 4)
>
Troemner Stainless Steel Precision Analytical Mass Sets
>
Troemner Class 1 Calibration Mass Sets with a NVLAP* Certificate of Calibration
Share
|
Precision ASTM Class 1 calibration mass set with an ISO/IEC 17025:2005-accredited certificate, 200 g to 1 mg
EW-11957-53
$2,202.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Precision mass sets are supplied in a 5-2-2-1 series; weights beginning with a “2” are doubled in each decade and weights beginning with a “5” and “1” are single in each decade
Mass sets of 500 g and lower include a clear-top polycarbonate case
*Troemner is accredited by the National Voluntary Laboratory Accreditation Program for the specific scope of accreditation under Lab Code 105013-0
Specifications
Product Type
Calibration Mass
Class
Class 1
Mass (g)
200
Contents of set
1 mg, two 2 mg, 5 mg, 10 mg, two 20 mg, 50 mg, 1 g, two 2 g, 5 g, 10 g, two 20 g, 50 g, 100 g, 200 g
Calibrations
Qty
/
Price
Availability
Calibration Service For mass Set With Maximum Value Between 100 To 200 G; Astm 1-3
EW-17109-34
at $368.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title