United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-61600-03
Image is sample only. Check individual details.
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-61600-03
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Panel Meters
>
Loop Leader Series Loop-Powered Rate/Totalizers
Share
|
Precision Digital PD689-0K1 Process meter, hazardous area
EW-61600-03
$404.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Intrinsically safe model for use in just about any hazardous area.
5-digit backlit display with bargraph
Open collector alarm or pulse output
HART protocol transparent
The loop-powered rate/totalizers can display rate up to five digits and total up to eight digits using the total overflow feature. The display can be programmed to automatically or manually toggle between rate and total. Standard features include a full 5-digit backlit display with bargraph, on-screen custom engineering units, and max/min display capabilities.
Square root and ptrogrammable exponent functions are standard features on these meters. These indicators can display flow rate and total from a DP transmitter or open channel flow in weirs and flumes. Programming is easy through the fron panel buttons, with no pots or jumpers to deal with, and can be scaled without a signal source.
The Loop Leader Series meters go just about anywhere. The PD689, with FM, CSA and ATEX certifications, can be installed in just about any hazardous area.
Specifications
Input
4 to 20 mA
Display
5-digit LCD, 0.6" (15.2 mm) high; 20-segment bar graph
Operating conditions
-4 to 149°F (-20 to 65°C)
Enclosure
1/8 DIN, high-impact plastic, UL 94V-0
Dimensions
4.68"W x 2.45"H x 3.79"D (119 x 62 x 96 mm)
Manufacturer number
PD689-0K1
Model
PD689-0K1
Technical Resources
Related technical articles (Roll over for summary)
Process/Panel Meter Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title