Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Primary High-Efficiency Filter w/ Carbon Module for Ductless System
Item# EW-33703-42
$320.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 5 days.
Qty
    Easily install or change filters in just seconds. Filter packages designed to suit your specific working conditions
    High-Efficiency Filters captures smok and dust — ideal if you mix dry chemicals, sand, grind, or do light welding. If you use solvents or paint in the same process, choose a combination high-efficiency filter with refillable 2" thick carbon module. Filter is 95% efficient.
    HEPA Filters capture soldering fumes and vapors that contain lead. If you use solvents or paint in the same process, choose a combination HEPA filter with refillable 2" thick carbon module. Filter is 99.97% efficient.
    Activated Carbon Filter captures volatile solvents, touch-up paint fumes, and other noxious odors. The primary
    Product Type Fume Hood Filters
    Qty/ea 1
    Brand Air Impurities Removal Systems
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456