log in or register
Primary Merv 14 Cell Filter
EW-33704-91
$186.00 USD/EACH
Usually ships in 8 days.
Qty
  • Multi stage filtration
  • Durable cabinet construction
  • Convenient handle and casters provide easy mobility
  • Highly efficient, yet low-cost replacement filters
  • Quiet operation
  • Long life blower motor
  • Adjustable extractor arm with fire-retardant properties
Portable air cleaning system is designed for direct source-capture removal of smoke, dust, fumes, and other airborne contaminants from a wide variety of industrial and other applications. Use to improve the air quality in your facility by effectively removing airborne contaminants beforre they enter the user''s breathing zone. Easily place adjustable extractor with aluminum hood where you need work done.
What's Included:
Four filters, three that are replaceable
Brand AIR IMPURITIES REMOVAL SYSTEM
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title