Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Pro/Clean Frock, Small

EW-33683-40
$177.00 USD/CASE of 30

Usually ships in 14 days.
Qty
    Pro/Clean®apparel is made of continuous fibers that have good stretchability, and a microporous film layer on the outside that reduces particle counts. This lets the fabric breathe while providing a protective barrier. All Pro/Clean® apparel is made with serged seams—a basic stitched seam that is appropriate for exposure to nonhazardous substances.Pro/Clean® Frocks have a zipper front and elastic wrists. Serged seams. Case of 30.
    Product Type Lab Coats
    Description Frock
    Size Small
    Color White
    seam Serged
    Material Polyethylene
    Compliance ISO Class 5 (formerly Class 100)
    Quantity 30/cs
    Manufacturer number PC271SWHSM00300B
    Brand Pro/Clean
    Model PC271SWHSM00300B





    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title