Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Programmable Fluid Bed Dryer, 110 VAC, 50/60 Hz

EW-02658-40
$10,594.00 USD/EACH

Usually ships in 7 days.
Qty

  • Quickly and evenly dry particles as small as 5 um
  • High reproducibility is ensured with accurately controlled and uniform drying process
  • Faster and gentler than conventional drying cabinets—ideal for food products, polymers, soils, chemicals, pharmaceuticals, minerals, and waste materials.
  • Variety of accessories provide easy customization for your drying applications
For a complete system, you need a fluid bed dryer, a sealed or non-sealed tub assembly, and filter top caps or filter bags. Optional accessories include low-density classifier assembly, temperature/humidity probe, pulser unit, software, and RS-232 cable.
This compact benchtop unit uses a powerful air delivery system and heater to rapidly remove moisture or solvents from samples with up to 80% moisture content. The unit ensures thorough drying, even with fine or irregularly-shaped samples. Store a program with up to 16 steps to optimize airflow rates, temperature, and drying time. For greater control, order the optional software and RS-232 cable to interface with a computer. The software allows you to program any number of steps, ending each step at a specific time, humidity, or outlet temperature.
Dimensions 12-5/8"W x 10-1/4"H x 20"D
Sample volume 2L, 5L, or 1000 mL (four 250 mL samples), depending on tub assembly used. Max weight 11 lbs (5 kg)
Power 110 VAC, 60 Hz, 23.6 A

Accessories
Qty/Price
Availability

at $4,186.00 USD/EACH

Usually ships in 7 days.

at $2,262.00 USD/EACH

Usually ships in 7 days.

at $1,722.00 USD/EACH

Usually ships in 7 days.

at $1,410.00 USD/EACH

Usually ships in 7 days.

at $519.00 USD/EACH

Usually ships in 7 days.

at $1,711.00 USD/EACH

Call 1-888-358-4705 for availability.

at $1,576.00 USD/EACH

Call 1-888-358-4705 for availability.

at $2,183.00 USD/EACH

Call 1-888-358-4705 for availability.

at $2,146.00 USD/EACH

Call 1-888-358-4705 for availability.

at $3,152.00 USD/EACH

Call 1-888-358-4705 for availability.

at $7,421.00 USD/EACH

Call 1-888-358-4705 for availability.
Build a complete system




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title