United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 25
of
68 >
EW-76803-38
Image is for illustrative purposes only. Please refer to product description
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-76803-38
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Positive Displacement Pumps
>
Progressing Cavity Pumps
>
Seepex Progressing Cavity Pumps
Share
|
Progressing cavity pump, Stainless steel body and Viton stator construction, 3 HP AC motor, 45 PSI max pressure, 46 GPM max flow
EW-76803-38
$3,432.00 USD/EACH
Usually ships in 10 days.
Qty
:
Add to Cart
Save to shopping list
Excellent chemical resistance, pulseless fl ow, and low shear
Easily move a variety of polymers and chemicals, and extremely adaptable to a wide range of industries
Gentle low-shear fluid transfer capable of pumping small particles without damaging product or pump
These pumps are of two main configurations: iron bodies with NBR stators and stainless steel (SS) bodies with Viton® stators. The two material combinations are ideal for many different industries including food & beverage, environmental, chemical & biochemical, pulp & paper, oil, gas & petrochemicals, agriculture, construction, biogas, ceramics, pharmaceutical and cosmetics . . . just to name a few!
The unique design of the rotor/stator combination provides many benefits—the smaller rotor cross section results in reduced thrust loads, the universal joint design offers improved NPSHR and low component wear, and the plug-in shaft connection allows for quick part change out. Listed below are the three most commonly asked-for drive configurations. The first is a standard AC motor (STD), the
second is a standard AC motor with an integrated controller to facilitate your changing requirements (IVD), and the third is the Spaggiari drive system (SPG).
Specifications
Product Type
Progressing Cavity Pumps
CE Compliance
Yes
Brand
Seepex
Manufacturer number
W05NA4H0BD2036
Model
W05NA4H0BD3034
Technical Resources
Related technical articles (Roll over for summary)
Positive Displacement Pump Types
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title