log in or register
Protector Premier, 6', Two Services and Duplex, No Blower, 115V, 60Hz
EW-09102-74
$9,385.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Excellent chemical resistance
Labconco Protector Premier Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general-purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.
These by-pass hoods feature the largest unobstructed sightline available—37.5” from work surface to header panel. Other features include Eco-Foil air foil that corrects natural turbulence while keeping cords out of the way, dry powder-coated steel exterior, safety glass sash, and molded fiberglass exhaust collar.
Hoods are available with a built-in standard fiberglass blower, an explosion-proof fiberglass blower, or without a blower (remote blower sold separately). Standard hoods include T8 fluorescent lighting. Explosion-proof (EP) models include incandescent lighting (bulb not included). EP blowers are UL-listed for Class I, Group D and Class II, Groups E, F and G. All models are ETL listed. 230V models are CE listed.
Additional service fixtures can be added; contact a Cole-Parmer Application Specialist.
Product Type Ducted Fume Hood
Exterior Dimensions (" W) 72
Exterior Dimensions (" H) 59
Exterior Dimensions (" D) 31.7
Sash opening (in.) 37.5
Blower (hp) 0.5
Blower (type) None
Power (VAC) 115
Power (Hz) 60
Hood size 72"
Fixtures Two
Ductwork size 12"
Lights Flourescent
Switches Two
Shipping weight 495 lb (225 kg)
Accessories
Qty/Price
Availability
at $3,029.00 USD/EACH
Call 1-888-358-4705 for availability.
at $3,101.00 USD/EACH
Call 1-888-358-4705 for availability.
at $2,732.00 USD/EACH
Call 1-888-358-4705 for availability.
at $366.00 USD/EACH
In stock.
at $554.00 USD/EACH
In stock.
at $342.00 USD/EACH
In stock.
at $569.00 USD/EACH
In stock.
at $377.00 USD/EACH
Usually ships in 8 days.
at $668.00 USD/EACH
Usually ships in 9 days.
1/2 hp Blower
EW-33704-80
at $1,242.00 USD/EACH
Usually ships in 14 days.
3/4 hp Blower
EW-33704-85
at $1,835.00 USD/EACH
Call 1-888-358-4705 for availability.
Related products
Qty/Price
Availability
at $862.00 USD/EACH
Usually ships in 35 days.
at $2,594.00 USD/EACH
Usually ships in 35 days.
at $5,454.00 USD/EACH
Usually ships in 35 days.
at $1,486.00 USD/EACH
Usually ships in 42 days.
at $1,820.00 USD/EACH
Usually ships in 42 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title