United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
6 >
EW-07164-52
Pump, Vacuum Pump, Rotary Vane, 6.0 CFM, 115/220 VAC, 2 Stage
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07164-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Vacuum Pumps
>
Oil Lubricated Vacuum Pumps
>
Rotary Vane Vacuum/Compressor Pumps
>
Economical Rotary Vane Pumps
Share
|
Pump, Vacuum Pump, Rotary Vane, 6.0 CFM, 115/220 VAC, 2 Stage
EW-07164-52
$415.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Compact design ideal for portable applications
Designed to minimize vibration
Dual Voltage
Isolation Valve and Gas ballast protects pump and system
Whether you work in the lab, manufacturing floor, or HVAC industry this is the pump for your applications. The lightweight design and rugged construction lend itself to even the most demanding environments. Dual-stage pumps feature two pump units connected in series which are driven from the same shaft to create vacuum levels as low as 5 x 10
-2
torr.
Large finned die-cast aluminum oil reservoir extends time between oil changes and helps limit overheating. All models have an isolation valve to prevent oil suck back into the system, angled low profile oil drain for fast and complete oil changes and large sight glass on oil reservoir to monitor oil level and condition. Center-balanced handle make transportation easy and clean with no oil spills.
Specifications
Max vacuum
5 x 10
-2
Free-air capacity
6 CFM (170 LPM)
Port size
1/4", 3/8" flare and 1/2 ACME (auto)
Max temperature
110°F (43°C)
hp
1/2
Dimensions
14.5" x 6.875" x 10.75" (37 x 17.5 x 27 cm)
Gas ballast
Yes
Tank size
15.2 ounces (0.45 L) oil capacity
Amps
5.4 A
Power
110/220 VAC
Stages
2
Description
Yellow color
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Basics of Cartridge Filtration
A Vacuum Pump That Survives Freeze Dryer Applications
Diaphragm Vacuum Pump Buyer's Guide
Show All...
Vacuum/Compressor Pump Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title