Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Pyrex 3900 Arsine Generator, 125 mL, 1/ea
Pyrex  |   Item# EW-34530-20
$133.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Qty
    Arsine generator determines inorganic arsenic as per diethyldithiocarbamate photometric method, which meets EPA, USP, and ACS specifications. Includes 125-mL flask, absorber, arsine scrubber, and KECK® ball-joint clip.
    Product Type Distillation Glassware
    Description Arsine generator
    Qty/ea 1
    Brand Pyrex
    Model 3900
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456