Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Pyrex 3915-C PYREX Accelerated One-Step Glass Extractor/Concentrator System
Pyrex  |   Mfr# 3915-C  |   Item# EW-34510-00
$1,771.00 USD/CASE of 1
Are you a reseller or buying in bulk? Call us today.
Usually ships in 14 days.
Qty
    Accelerated One-Step™ modular extractor/concentrator performs extraction, concentration, and drying—all in one apparatus! Features PTFE hydrophobic membranes for speedy drying. Meets EPA and CLP standards. Includes extractor cup and body, Snyder sidearm, membrane assembly, and connectors. Order concentrator tube and condenser separately below to fit your specific application.
    Product Type Extraction Glassware
    Qty/cs 1
    Brand Pyrex
    Manufacturer number 3915-C
    Accessories
    Qty/Price
    Availability
    at $195.00 USD/CASE of 1
    Available for order
    or call 1-888-358-4705 to order
    at $288.00 USD/CASE of 1
    Available for order
    or call 1-888-358-4705 to order
    at $198.00 USD/CASE of 1
    Usually ships in 10 days.
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456