United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 9
of
12 >
Representative photo only
RII Safety Coated Vertical Rotary Evaporator System 115V
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-38605-40
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Evaporators
>
Rotary Evaporator Systems
>
Buchi Rotary Evaporators and Accessories
>
Buchi Rotavapor RII Rotary Evaporators
Share
|
RII Safety-Coated Vertical Rotary Evaporator System, 115V
EW-38605-40
$4,770.00 USD/EACH
Usually ships in 3 days.
Qty
:
Add to Cart
Save to shopping list
Durability and reliability with simple operation
Small footprint to maximize your lab space
Choice of three glassware configurations for application versatility
Glassware available with protective safety coating to minimize breakage
Designed specifically to meet standard distillation needs in the educational laboratory, these economically priced models feature analog controls for both rotation and bath temperature. Set rotation speed from 20 to 280 rpm. Heating bath features corrosion-resistant coating and operates from 20 to 95°C; quick-release electrical connection makes it particularly easy to remove for fluid removal or addition. Manual lift allows the evaporator to be raised or lowered by hand, making operation quick and safe.
Choose from three condenser assemblies for application versatility. Use the diagonal condenser assembly for standard distillations and where lab space limits height. The vertical condenser is used for standard distillations but takes up less bench space than the diagonal unit. The cold-trap condenser is
ideal for applications without cold water circulation.
Select from systems with standard glassware or glassware with Plastic + Glas
®
coating, which protects you against broken glass and dangerous reagents due to broken glass. Safety coating resists most solvents and acids, and is available on all glassware except evaporating flask.
Note:
Direct exposure of Plastic + Glas glassware to chlorinated solvents and aggressive reagents should be avoided. After extensive use, coating on glassware may wear and loosen from surface, increasing risk of injury.
What's Included:
water bath, condenser set, evaporating and receiving flasks (each 1 L), and 6-ft power cord.
Specifications
Bath temperature
20 to 95°C
Bath Capacity
4 L
Temp control accuracy
+/- 5C
Power (Hz)
60
Dimensions
21-21/32"W x 22-41/64"H x 16-11/32"D (55 x 57.5 x 41.5 cm)
Rotation speed range
20 to 280 rpm
Evaporation flask size
50 mL to 4 L
Watts
1300
Vacuum seal
PTFE
Joint size
24/40
Lift mechanism
Manual
Condenser type
Vertical Evaporators
Glass type
Plastic and Glas coated
CE Compliance
Yes
Brand
Buchi
Power (VAC)
115
Manufacturer number
2422V1
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title