dummy
EW-86551-22
Call 1-888-358-4705 for availability.
Qty:
- Versatile, easy-to-use monitors can be used in both laboratory and field applications
Rugged monitors are constructed with a NEMA-4X polycarbonate case that is corrosion resistant and splash-proof to withstand the rigors of every day use. Large, analog bar graph display is readable at a glance without complicated programming or electronicsjust power up and go. Monitors operate on battery or AC power source for portable or stationary use. Model 86551-20 features a 2'' pancake probe to read alpha, beta, and gamma radiationideal for clothing, benchtop, or hand surface readings. Model 86551-22 features a 3'' scintillation probe to read gamma and X-ray radiationsuitable for entryway or area monitoring.
What's Included:
probe, probe holder, two 9 V transitor batteries, and 100-125 VAC adapter.
| Display |
multirange analog bar graph |
| Function |
adjustable sensitivity, audible/visible alarms |
| Detector |
3" scintillation |
| Window |
NaI (T1) crystal and 5-mil thick window |
| Dimensions |
9" x 5 1/2" x 4" (22.9 x 14.0 x 10.2 cm), meter only |
| Output |
0 to 1 volt DC full-scale |
at $25.60 USD/PACKAGE of 4
In stock.
Technical Resources
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334