United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-05465-60
Raven Coretaker Polycarbonate Sludge Sampler, 16-ft
Click to enlarge
More Product Details
Specifications
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-05465-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Samplers
>
Water and Industrial Liquid Sampling
>
Sludge Sampling and Settled Solids Measurement
>
Raven Environmental Products Coretaker for Sludge Blanket Measurements
Share
|
Raven Coretaker Polycarbonate Sludge Sampler, 16-ft
EW-05465-60
$108.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for direct measurement of sludge blanket thickness by core sample extraction
Lifetime, no breakage gaurantee
Floats for easy retrieval
No threaded couplersuses Slip-Loc quick-disconnect design
Two, rigid 8-ft lengths of clear, 1-1/4”dia polycarbonate tubing will not turn cloudy from sunlight or become brittle in cold weather like acrylic sludge samplers. The design offers a Slip-Loc quick-disconnect coupler and twin double o-ring seals to ensure a rugged, liquid-tight connection. Coretaker features a check valve for sample release; soft foam, floating lanyard handle; and pre-numbered yellow 1-ft markers.
Specifications
Dimensions
1-1/4" dia x 16-ft L
Manufacturer number
B-10104-16-REDESIGNED
Model
B-10104-16-REDESIGNED
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
E/S Portable Sampler
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title