United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
10 >
EW-68515-99
Sample image. Check individual details.
Click to enlarge
More Product Details
Specifications
Accessories
Reconditioned & Like New
Technical Resources
Instruction manual
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68515-99
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Automation Control
>
Red Lion Data Station Plus Operator Interface with Protocol Translator and Plant Floor Marquees
Share
|
Red Lion Data Station Plus DSPSX Operator Interface and Protocol Converter with email/text message on alarm feature
EW-68515-99
$930.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
A nexus for industrial data collection and management
Converts numerous protocols simultaneously
Over 200 built-in drivers allow easy data mapping to PLCs, PCs, drives and SCADA systems
DSPSX version comes with standard web server, data logging (via CSV file format) and alarm e_mail/text messaging
Supports up to nine protocols simultaneously (with expansion card)
DIN rail mounting
Operating termperature 32 to 122°F (0 to 50°C)
The Data Station Plus is designed to act as a nexus for industrial data collection and management. With three built-in serial ports and a 10 Base-T/100 Base-TX Ethernet port, the unit performs protocol conversion, allowing disparate devices to communicate seamlessly with one another. The Data Station Plus functions both as a data management platform and as a PFM host.
The Ethernet port supports up to four protocols simultaneously so even Ethernet-to-Ethernet protocol conversion may be performed. The DSPSX version also has a built-in web server, providing worldwide access to data logs and virtual HMI.
The CompactFlash card allows new configuration files to be loaded into the Data Station Plus or use to data log critical information. The USB port may be used for fast file downloads, or to mount the Data Station’s CompactFlash card as an external drive to your PC.
The DSPSX version also features e-mailing/text messaging capabilities that allow alarms to be forwarded to any cell phone or network PC.
What's Included:
Hardware packet for mounting unit into panel.
Specifications
Product Type
Operator Interfaces
Model
DSPSX000
Dimensions
8.03"W x 5.91"H x 1.89"D (204 x 150 x 48 mm)
Brand
Red Lion
Accessories
Qty
/
Price
Availability
Red Lion Data Station Plus XCCN0000 CANopen Option Card
EW-68516-15
at
$203.00 USD/EACH
In stock.
Add to Cart
Red Lion Data Station Plus XCDN0000 DeviceNet Option Card
EW-68516-16
at
$407.00 USD/EACH
In stock.
Add to Cart
Red Lion Data Station Plus XCPBDP00 PROFIBUS Option Card
EW-68516-17
at
$407.00 USD/EACH
In stock.
Add to Cart
Red Lion Data Station Plus XCRS0000 RS232/485 Option Card
EW-68516-18
at
$142.00 USD/EACH
In stock.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
RECOND 68515-99
EW
-
00315DJ
at
$605.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Data Station Plus-DSP Model Instruction Manual.pdf
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title