United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
10 >
EW-44200-00
Refrigerator/freezers (two doors)
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-44200-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Refrigerators
>
Refrigerator and Freezer Combination
>
Safety Refrigerators/Freezers
>
Thermo Scientific General-Purpose and Safety Refrigerator/Freezers
Share
|
Refrigerator/freezers; two doors; Description, Explosion Proof; 120V
EW-44200-00
$4,030.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Combination storage for biological materials or chemicals
CFC-free insulation and coolant
Explosion-Proof Models
protect against explosions both inside and outside of the unit. All motors, switches, and thermostats prohibit arcing that can ignite flammable air-vapor mixtures. They meet Class 1, Group C and D requirements for hazardous environments. Explosion-proof units must be hard-wired to voltage source.<
Specifications
Product Type
Explosion Proof Combos
Capacity (cu ft)
8.6/2.4
Capacity (Liters)
244/68
Dimensions (overall)
23-1/2" x 60" x 29-1/4"
Min Temp - Refrigerator (º C)
-1
Min Temp - Refrigerator (º F)
30
Max Temp - Refrigerator (º C)
8
Max Temp - Refrigerator (º F)
46
Min Temp - Freezer (º C)
-20
Min Temp - Freezer (º F)
-4
Max Temp - Freezer (º C)
-12
Max Temp - Freezer (º F)
-10
No. of doors
2
No. of shelves
3 cabinet, 4 door
Power (Hz)
60
Power (watts)
230
Brand
Thermo Scientific
Manufacturer number
3551-DB
Power (VAC)
120
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title