United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
10 >
Representative photo only
Regenerative Blower 74 92 cfm 3 phase
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
More info
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07047-35
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Air Compressors
>
Regenerative Blowers
>
Regenerative Blowers
Share
|
Regenerative Blower, 74/92 cfm, 3 phase
EW-07047-35
$1,140.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Quietly recirculate air at up to 420 cfm
TEFC motor mounted
Use in any orientation to meet your system requirements
Perfect for pond or plate bath aeration, wastewater treatment, and trim and dunnage removal
Cord and plug not included.
*Performance is decreased to 5/6 when operating at 50 Hz
Specifications
Product Type
Regenerative Blowers
Free air capacity
74/92 cfm
hp
2/3; 1
Amps
2.6-3.3/1.3-1.4 at 190-220/380/415 VAC; 3.4-3.2/1.6 at 208-230/460 VAC
Thread size
1.5" NPT(F)
Wetted parts
Cast Iron, Cast Aluminum
Port size
1.5" NPT(F)
Duty cycle
Continuous
Dimensions
12.4375" L x 11.25" W x 11.75" H
Power (VAC)
190 to 460
Accessories
Qty
/
Price
Availability
Inlet filter for Regenerative Blowers
EW-07047-12
at
$45.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Regenerative Blower Performance Curves
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title