United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
Representative photo only
Reichert Refractometer 0 to 50 Brix
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-81007-52
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17105-00
NIST-Traceable Calibration; Refractometer
$75.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Refractometers
>
Handheld Manual Refractometers
>
Reichert Rhino Series Brix Refractometers
Share
|
Reichert Refractometer, 0 to 50% Brix
EW-81007-52
$174.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Feature automatic temperature compensation—no correction tables or temperature measurement required
Water-resistant IP67-rated design is ideal for stringent environments
Rugged, impact-resistant Xenoy
®
polycarbonate housing withstands repeated drops
These refractometers offer exceptional optical design and improved image resolution with high contrast for easy viewing of scales. All reflection-coated glass optical components, including a 7x eyepiece, have been developed to improve illumination and shadowline contrast. Black housing improves image quality by reducing stray light. Optics and instrument scales are sealed inside, safe from harsh external conditions. Measure concentration of fruits, beverages, and canned goods; total solid content of syrups, vinegar, and sauces; or check the quality of vegetable, fruit, and grass tissues.
What's Included:
sample pipette and carrying case.
Specifications
Product Type
Refractometer (Analog)
Brix scale range (Minimum)
0
Brix scale range (Maximum)
50
Resolution
0.25% Brix
Temp compensation
0 to 104°F (-18 to 40°C)
Accuracy
±0.25% Brix
Brand
Reichert
Model
137531LO
Manufacturer number
137531LO
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Refractometer
EW-17105-00
at $75.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Testing Refractive Index of Candy
Refractometers
Brix Scale (sucrose)
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Electron zap turns flimsy plastic into sturdy shrink wrap
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title