Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Replacement 100 W Mercury Lamp f/ Epifluorescent Microscope
Meiji  |   Mfr# BA005   |   Item# EW-48924-66
$210.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 4 days.
Qty
  • Semi Apo Plan objectives provide bright, high-contrast images, with UV transmission and color correction
Epi-fluorescent microscope is designed for use with specimens that self fluoresce, or that use a fluorescent dye to bring out particular properties of the specimen. Ideal for use in cell biology and laboratory research. Modular six-position epi-fluorescent illuminator features six blank positions for filter cubes; basic blue, green, and UV-excitation filter sets mounted in filter cubes are included. Lamp house uses a 100 W mercury arc lamp.
Microscope features include siedentopf head for adjustability, diopter adjustment on one eyepiece, ceramic-coated stage, Abbe condenser with adjustable field iris, bottom halogen illumination for transmitted light use, and ergonomically placed focus controls.
What's Included:
mercury lamp power supply; blue, green, and UV-excitation filter sets; and a dust cover.
Product Type Replacement bulb
Manufacturer number BA005
Brand Meiji
Qty/ea 1
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456