log in or register
Replacement free chlorine test kit, for Oakton colorimeters, 100 tests/kit.
EW-35645-64
$25.00 USD/EACH
  • Reagents for Free and Total Chlorine, Ozone and Chlorine Dioxide
  • High quality powder reagents for exceptaonal accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • 100 tests per pack
Product Type Accessory
Tests/kit 100
Manufacturer number 94X377001
Brand Oakton
Technical Resources
  • MSDS 1 (135 B)
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title