Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Rexyn* I-300 (H-OH) Beads (Analytical Grade/Certified) (500g)

Mfr# R276500  |   Item# EW-88079-97

$310.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.

Usually ships in 2 days.

  • Mesh Size (wet, U.S. Standard Sieve) - To pass test (16-50 mesh)
  • Total Exchange Capacity (wet volume) - ≥0.55mEq/mL
  • Total Exchange Capacity (dry basis) - ≥1.55mEq/g
  • Active Working Density (dry basis) - 340g/L
  • Moisture Content - 50-60%

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Reagents
Purity grade Certified
Form Beads
Packaging Amber Glass
Storage symbol Gray
Manufacturer number R276500

Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews