United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
6 >
Representative photo only
Rice Lake Class 1 Calibration Weight Set 100 g 1 mg
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10151-11
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17109-10
Calibration Service For mass Set With Maximum Value Between 100 To 200 G; Astm 0-3
$997.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Calibration Masses
>
Rice Lake Calibration Masses
>
Rice Lake Class 1 Precision Mass Sets with Certificate of Calibration
Share
|
Rice Lake Class 1 Calibration Weight Set 100 g - 1 mg
EW-10151-11
$1,775.00 USD/EACH
Usually ships in 9 days.
Qty
:
Add to Cart
Save to shopping list
Ensure balance accuracy with calibration masses from Rice Lake
Weights conform to NIST, ASTM or OIML (unlike internal calibration masses)
4 day turnaround time on new masses and mass recalibrations
Individual masses and sets from Class 0 through Class 4
Electronic balance calibration masses
Specialty masses and custom designs
Succinct traceable calibration certificates
Ensure balance accuracy with calibration masses from Rice Lake. Dedicated to excellence and service, Rice Lake offers a 4-day turnaround on masses and calibrations.
Masses are made of polished stainless steel and include a glove, case and lifter unless otherwise indicated. As the leader in service, selection and speed – Rice Lake stands behind our offerings with a commitment beyond measurement.
Specifications
Product Type
Calibration Mass
Class
Class 1
Mass Set
100 g - 0.001 g weight set
Model
11897CR
Manufacturer number
11897CR
Calibrations
Qty
/
Price
Availability
Calibration Service For mass Set With Maximum Value Between 100 To 200 G; Astm 0-3
EW-17109-10
at $997.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title