Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Rochester Pean Forceps, Dissecting-grade, Straight

EW-10921-40
$26.25 USD/EACH

Call 1-888-358-4705 for availability.
Qty
    Dissection-grade instruments Vantage® brand by Miltex, are manufactured from stainless steel, hardened, tempered,and polished for a high-priced finish without the added cost. These instruments are well-suited for less critical applications with tight budgets such as necropsies.
    Product Type Forceps
    Description Rochester Pean Forceps
    Dimensions (" L) 7.25
    Material Stainless Steel
    End type Straight
    Grade Dissecting
    Brand Miltex
    Manufacturer number V97-122





    Technical Resources
     
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title